Would you like to log in again?
Educational Diffraction Slide
Supplier: EISCO SCIENTIFIC LLC
PH0625
470230-222EA 115.8 USD InStock
470230-222
Educational Diffraction Slide
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrates the principle of diffraction grating.
  • For demonstration principle of diffraction grating
  • Comprises a card mount 90 x 40 mm with three 16 x 9 apertures
  • Gratings are 100 lines/mm, 300 lines/mm, 600 lines/mm
  • Include protected cover glasses
For demonstrating the principle of the diffraction grating and the way in which its properties are dependent upon the number of lines per unit length. Comprises a card mount 90 x 40 mm approximately with three 16 x 9 mm apertures, each containing a diffraction grating replica. The grating are of 100 lines / mm, 300 lines / mm and 600 lines /mm respectively and protected cover glasses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty