Would you like to log in again?
Power Pack
Supplier: EISCO SCIENTIFIC LLC
PH0676A
470230-232EA 127.5 USD InStock
470230-232
Power Pack
Educational Materials Physics Educational Materials Optics Learning Activities
Low voltage power supply that can be used as a battery eliminator.
  • Switchable between 6 V or 12 V, AC or DC (at 2 Amps, not adjustable)
  • Banana plug terminals
  • Perfect for use with optical benches or other physics equipment
  • Can be used as a high powered battery eliminator
Power pack is a low voltage power supply that operates on 110 V power and outputs 6 or 12 V at 2 Amp either AC or DC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty