Would you like to log in again?
Wave Apparatus, Plastic
Supplier: EISCO SCIENTIFIC LLC
PH0712N8
470230-242EA 101.75 USD InStock
470230-242
Wave Apparatus, Plastic
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Plastic wave apparatus ideal for studying wave behavior.
  • Demonstrate longitudinal and transverse waves
This advanced apparatus demonstrates both longitudinal and transverse wave motions. Consisting of a number of eccentric discs supporting a series of metal rods revolving on a handle, transverse waves are obtained. Longitudinal waves are obtained from the bent rods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty