Would you like to log in again?
Sympathetic Tuning Fork on Wooden Base
Supplier: EISCO SCIENTIFIC LLC
PH0742A
470230-252PK 43.45 USD InStock
470230-252
Sympathetic Tuning Fork on Wooden Base
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Steel tuning forks for physics experiments.
  • Used to hear dissonance
  • One fork adjustable
  • Comes with mallet and bases
For demonstrating the production of beat frequencies. Comprises two nickel plated forks A (440 Hz ±5%) mounted on individual boxes. One fork is provided with adjustable mass by means of which frequency may be shifted from the nominal 440 Hz .When both forks are sounded a clearly audible 'beat' is produced, its rate depending upon the difference in frequency between the forks. (2) Tuning forks with bases, (1) Mallet.
Note: These are designed for physics experiments to create beats and are often not exactly 440 Hz (usually 436 - 444 Hz). They can be adjusted to create beats within each other or to match each other.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty