Would you like to log in again?
Organ Pipe
Supplier: EISCO SCIENTIFIC LLC
PH0759
470230-256EA 57.9 USD InStock
470230-256
Organ Pipe
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Polished wood organ pipe for investigating beat phenomenon.
  • Polished wooden organ pipe
  • Fully extends to 34.5"
  • Marked with chromatic scale from C(512) to (1024)
  • Ideal for investigating beat phenomenon using a second pipe
Labs polished wooden organ pipe with movable piston is marked with the chromatic scale from C(512) to (1024). It has a fully extended length of 34.5" and works well for investigating the phenomenon of beats in conjunction with a second pipe of the same type.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty