Would you like to log in again?
Tryptic Soy Broth, Biomerieux
Supplier: bioMérieux
P2760-RS P2310 P2300
95061-484CS 317.93 USD InStock
95061-484 95061-482 95061-504
Tryptic Soy Broth, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Broth is a general purpose medium used for the cultivation of fastidious and non-fastidious microorganisms.
  • General purpose medium
  • Multiple sizes available
Upon receipt, store bottles and tubes according to the package label in an upright position away from direct light.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty