Would you like to log in again?
Tryptic Soy Agar with Lecithin and Polysorbate 80, Biomerieux
Supplier: bioMérieux
P2810
95061-512CS 504.96 USD InStock
95061-512
Tryptic Soy Agar with Lecithin and Polysorbate 80, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Agar with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
  • For isolation and cultivation of microorganisms from surfaces sanitized with quaternary ammonium compounds
  • 10 x 500 mL
Upon receipt, store plates at 2 – 8 °C away from direct light in an inverted position. Store bottles according to the package label in an upright position. Store plates in original outer wrapping until ready to use. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty