Would you like to log in again?
UUVBioTAG™, Microbiologics
Supplier: Microbiologics
UVBioTAG™
01248UV-V 01248UV-S 01249UV-V 01249UV-S 01250UV-S 01250UV-V
76124-798PK 481.62 USD InStock
76124-798 76124-800 76124-802 76124-804 76124-806 76124-808
UUVBioTAG™, Microbiologics
Control reagents
UV-BioTAG, our line of reference strains containing green fluorescent protein (GFP) markers, makes it easy to distinguish standard microorganism strains from laboratory control strains.
  • Fluoresces under UV light to distinguish QC strains from other possible contaminants.
  • Instant dissolve pellet reduces preparation time.
  • Ready-to-use format saves time and money.
  • Refrigerated storage is easy and economical.
  • Online Certificate of Analysis provides detailed strain information.
  • Traceability to reference cultures ensures authenticity.
Optimized for highly visible fluorescence, UV-BioTAG strains provide better stability because the GFP marker is integrated into the chromosome rather than the plasmid. With UV-BioTAG, you can quickly and reliably establish whether a positive test result can be traced back to a control strain cross-contamination issue, or if it is from another source.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty