Would you like to log in again?
SP Wilmad-LabGlass Glass Rod Stirrer Shafts, SP Industries
Supplier: SP Industries
LG-9500-102 LG-9500-106 LG-9500-100
14202-444EA 76.49 USD InStock
14202-444 14206-390 14209-834
SP Wilmad-LabGlass Glass Rod Stirrer Shafts, SP Industries
Stirrers Stirring Shafts
Made of borosilicate glass, these precision ground stirrer shafts are equipped with a side button for attachment of stirrer blades.
NOTE: Use blades LG-9505 through LG-9511T for LG-9500 shafts, ML-1770T blade for ML-1760 shaft, and LG-9510T-100 blade for ML-1765 shaft.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty