Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Wilmad-LabGlass Heavy Duty PTFE Stirrer Blades, SP Industries
Supplier: SP Industries
LG-9516T-108
14009-520EA 148.59 USD InStock
14009-520
SP Wilmad-LabGlass Heavy Duty PTFE Stirrer Blades, SP Industries
Stirrers Stirring Blades
These heavy duty PTFE stirrer blades are suited for stirring high viscosity liquids.
  • 3/16" thick blade
They are used with shafts that have a side button for blade attachment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty