Would you like to log in again?
SP Wilmad-LabGlass PTFE Banana Shape Stirrer Blades, SP Industries
Supplier: SP Industries
LG-9511T-128 LG-9511T-104 LG-9511T-102 LG-9511T-100
14201-726EA 73.94 USD InStock
14201-726 14211-466 14208-716 14211-348
SP Wilmad-LabGlass PTFE Banana Shape Stirrer Blades, SP Industries
Stirrers Stirring Blades
Made of 1/8" thick material, these PTFE stirrer blades are shaped so that bottom of the blade approaches more closely the actual inside contour of round bottom flasks.
  • Two styles
Offered in two styles for attachment to stirrer shaft; Type A with center opening, and Type B with center opening plus a number of other perforations for blade attachment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty