Would you like to log in again?
Proto® TorquePlus™ Impact Sockets, 1", 1" Drive, ORS Nasco
Supplier: ORS Nasco
577-10068 577-10050 577-10054 577-10029 577-10045 577-10041 577-10064 577-10062 577-10026 577-10047 577-10013 577-10023 577-10046 577-10014 577-10019 577-10043 577-10072 577-10036 577-10030 577-10058 577-10018 577-10032 577-10028 577-10035 577-10025 577-10022 577-10012 577-10015 577-10024 577-10039 577-10056 577-10021 577-10037 577-10044 577-10017 577-10020 577-10040 577-10031 577-10027 577-10033 577-10038 577-10052 577-10066 577-10042 577-10016 577-10048 577-10034
300023-290EA 335.89 USD InStock
300023-290 300023-192 300023-292 300023-270 300023-150 300023-250 300023-272 300023-174 300023-286 300023-242 300023-188 300023-144 300023-166 300023-222 300023-244 300023-200 300023-266 300023-300 300023-246 300023-204 300023-304 300091-598 300023-228 300023-208 300023-308 300091-594 300091-596 300023-182 300023-260 300023-282 300023-140 300023-162 300023-284 300023-252 300023-276 300023-232 300023-298 300023-178 300023-156 300023-278 300023-212 300023-256 300023-136 300023-312 300023-238 300023-316 300023-318
Proto® TorquePlus™ Impact Sockets, 1", 1" Drive, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty