Would you like to log in again?
HQ440d Benchtop Meter Package with ISENH4181 Ammonium ISE Electrode, Hach
Supplier: Hach
8507800
10199-512EA 5273.86 USD InStock
10199-512
HQ440d Benchtop Meter Package with ISENH4181 Ammonium ISE Electrode, Hach
Electrochemical Meters Ion Meters
Designed for water quality experts, the HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Flexible benchtop meter makes laboratory testing of critical water quality parameters more efficient
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements IntelliCAL™ smart probes store all calibrations in the probe
  • Enhance productivity with an ultrabright screen and large font size for easy to read results
  • Convenient kit includes everything needed to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous laboratory use. HQd meters feature a simple user interface that does not require manuals or training to operate.
Ordering information: Includes HQ440D Laboratory Multi Meter, Universal electrode Stand with electrode holder, ISENH4181 Ammonium Ion Selective Electrode with 1 m cable, Nitrogen Ammonia standard 500 ml solutions: 1 mg/L, 10 mg/L and 100 mg/L. Ammonium ISAB powder pillows pack (100 pcs), 4 Alkaline 1.5 V AA batteries, Power Supply Adapter, Power Cord, USB cable for data transfer, and Operating Documentation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty