Would you like to log in again?
GST SpinTrap Columns, Cytiva
Supplier: Cytiva
GST SpinTrap™
28-9523-59 28952359
97060-880PK 597.65 USD InStock
CA28952359 97060-880
GST SpinTrap Columns, Cytiva
Chromatography Columns
GST SpinTrap™ columns are designed for rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates. GST SpinTrap™ columns are suitable for purification of multiple samples in parallel, for example in screening experiments such as evaluation of chromatography media, growth temperature, culture density, and other variables to find optimal conditions for expression.
  • GST-tagged proteins can be recovered under mild elution conditions (10 mM glutathione), which preserves the functionality of the proteins
  • More than 90% purity can be obtained in a single step
Each column contains 50 μl of Glutathione Sepharose™ 4B, enough for purifying up to 500 μg of recombinant GST and each package contains 50 columns. For increased convenience, GST Buffer Kit contains stock solutions of the buffers needed for purification of GST-tagged proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty