Would you like to log in again?
SporView® Steam Self-Contained Biological Indicator, Crosstex/SPS Medical
Supplier: Crosstex
SCS-100 SCS-025
101101-804PK 216.13 USD InStock
101101-804 101101-802
SporView® Steam Self-Contained Biological Indicator, Crosstex/SPS Medical
Sterilization Indicators
Steam Self-Contained Indicators are used for biologically monitoring all steam sterilizers (gravity displacement and dynamic-air-removal operating at 121-135 °C).
  • For use with all steam sterilizers - reduces inventory and operator error
  • Inexpensive dry block incubator with built-in activator
  • Spore growth observable in as little as 2.5 to 24 hour final incubation
  • Media growth promotion validation per USP
  • Allows facilities to comply with industry guidelines for sterilizer monitoring
Each SporView Self-Contained Steam BI contains Geobacillus stearothermophilus spores and growth media along with a chemical indicator label printed with lot and expiration date. After processing, the SCBI is activated and then incubated at 55-60 °C for 24 hours. Media growth promotion has been validated for growth of G. stearothermophilus organism after exposure to a steam cycle of 132 °C for 20 minutes exposure.
Certifications: Certified by ANSI/AAMI standards; CSA standards and CDC guidelines.
Ordering information: A Starter Kit is available with 25 SCBIs with a pre-set incubator and record notebook.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty