Would you like to log in again?
Sephadex™ G-15 Gel Filtration Media, Cytiva
Supplier: Cytiva
Sephadex™
17-0020-01 17-0020-01
95016-750EA 449.74 USD InStock
CA95016-750L 95016-750
Sephadex™ G-15 Gel Filtration Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex G-15 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules >1500 molecular weight.
  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Suitable for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides larger than 1500 molecular weight
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
Sephadex is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex G-15 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large biomolecules.
Ordering information: Supplied in lab packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty