Would you like to log in again?
BBL™ Rappaport-Vassiliadis R10 Broth, BD Life Sciences
Supplier: BD Life Sciences
292915 292915
90008-109PK 433.23 USD InStock
90008-109 90008-108
BBL™ Rappaport-Vassiliadis R10 Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Rappaport-Vassiliadis R10 Broth has been involved in selective enriching of Salmonella from meat, feces, dairy products and sewage polluted water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty