Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Specimen Collection Kit, Capitol Vial®
Supplier: Capitol Vial
92023
76022-526CS 247.25 USD InStock
76022-526
Specimen Collection Kit, Capitol Vial®
Sample Containers General Purpose Containers
Split foil top specimen collection kit is used in performing sample collection and storage.
It is ideal for SAMHSA workplace testing.
Delivery information: Kit includes Beaker (236 mL) with temperature indicator strip, (2 to 45 mL), Generic dual pocket specimen bag, absorbent, foil top.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty