Would you like to log in again?
Weiler® Drive Arbor, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-07726 804-07727
300010-133EA 9.98 USD InStock
300010-133 300048-130
Weiler® Drive Arbor, ORS Nasco
Tool Sets
Drive arbor for mounting small diameter wheels into a collet or chuck
Use with brush diameters a maximum of 4".
It can reach speeds of 20,000 to 25,000rpm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty