Would you like to log in again?
Pyrolon® Plus 2 Disposable Flame Resistant Coverall with Hood and Boots, Lakeland Industries
Supplier: Lakeland Industries
7414B-2X 7414B-XL 7414B-MD 7414B-3X
10847-008CS 546.49 USD InStock
10847-008 10847-006 10847-004 10847-000
Pyrolon® Plus 2 Disposable Flame Resistant Coverall with Hood and Boots, Lakeland Industries
Disposable Garments
Disposable flame resistant coveralls with attached hood and boots protect primary flame resistant clothing from dirt, grease, and oil.
  • Disposable secondary flame resistant protection
  • Designed to be worn over primary flame resistant clothing
  • Oil and water replelant
  • Attached hood and boots
Disposable flame resistant coverall designed to keep primary flame resistant clothing free of dirt, grease, oil, and light liquid or spray. Coveralls have a zipper front with elastic wrists and attached hood and boots.
Certifications: Meets NFPA 2113 requirements for section 5.1.9.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty