Would you like to log in again?
Standards for Water Quality Analysis, Wako Chemicals
Supplier: Wako
132-07071 072-03421 077-01911 134-10581
100369-836EA 459 USD InStock
100369-836 100369-810 100369-688 100369-684
Standards for Water Quality Analysis, Wako Chemicals
Standards Environmental Analysis Standards
Moldy Odor Standards for Water Analysis.
The moldy odor produced by environmental pollution in rivers and lakes is due to metabolic products of various abnormally proliferated actinomyces and algae.
In particular, Geosmin and 2-Methylisoborneol (2-MIB) show a strong earthy odor (or moldy odor) with even an extremely trace, therefore they are considered to be most responsible for offensive odors in tap water.
Caution: Products are intended for laboratory research use only, and not to be used for drug, food or human.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty