Would you like to log in again?
Steelite™ Welted FW17, Safety Boots, Lace-Up, Portwest
Supplier: Portwest
Steelite™
FW17HOR47 FW17HOR44 FW17HOR48 FW17HOR42 FW17HOR45 FW17HOR46 FW17HOR41 FW17HOR39 FW17HOR40 FW17HOR43
76104-116EA 102.02 USD InStock
76104-116 76104-446 76104-444 76104-442 76104-440 76104-450 76104-438 76104-118 76104-448 76104-436
Steelite™ Welted FW17, Safety Boots, Lace-Up, Portwest
Shoes and Boots
Exceptional comfort and durability, this boot made from full grain Nubuck leather will protect with steel toecap, oil- and 300 °C heat-resistant rubber outsole.
  • Protective steel toecap
  • Full grain Nubuck leather
  • Goodyear welt construction
  • Slip-, fuel-, and oil-resistant rubber outsole
  • Heat-resistant outsole
The full grain leather upper provides a classic, long lasting style.
Color: Honey
Certifications: ASTM F2413-11.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty