Would you like to log in again?
illustra™ Sephadex® G-25 Grade DNA Clean Up, Cytiva
Supplier: Cytiva
illustra™ Sephadex®
17-0572-02 17-0572-02
95016-892EA 611.09 USD InStock
CA95016-892L 95016-892
illustra™ Sephadex® G-25 Grade DNA Clean Up, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For purification of DNA (10 bases in length) from small molecules by gel filtration.
  • Tested to ensure reproducibly high recovery of DNA
  • Used in MicroSpin G-25 Columns and NAP Columns
  • Enables self preparation of columns for nucleic acid purification
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty