Would you like to log in again?
CD™ Vials with PTFE Lined Screw Cap, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
CD™
CG-4915-01
89029-470EA 272.45 USD InStock
89029-470
CD™ Vials with PTFE Lined Screw Cap, Chemglass
Vials
These CD™ vials have a unique patented conical interior bottom permitting the contents to completely drain to the center bottom of the vial.
  • Less than 4 µl of dead volume
Maximum extraction can be accomplished using a syringe or micropipette.
Delivery information: Available complete with open top screw caps having a PTFE faced silicone septa or vials only.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty