Would you like to log in again?
S-Monovette® Blood Collecting Systems, Serum Gel (Clotting Activator), Sarstedt
Supplier: Sarstedt
S-Monovette®
04.1934.100
101433-392PK 20.95 USD InStock
101433-392
S-Monovette® Blood Collecting Systems, Serum Gel (Clotting Activator), Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® is a blood collection system that combines the aspiration technique and the vacuum technique.
  • Aspiration technique as a gentle technique for routine blood collection
  • Using the vacuum technique, a "fresh" vacuum is always available
  • Suitable for all ages, from young to old
S-Monovette® is suitable for all vein conditions and achieves an optimal sample quality for high quality results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty