Would you like to log in again?
Plater's Brushes, Weiler®
Supplier: Weiler
804-44119 804-44076 804-44118
300046-022EA 112.33 USD InStock
300046-022 300010-252 300010-253
Plater's Brushes, Weiler®
Brushes
Plater's brushes with brass or Tampico bristles and wooden handle.
  • Long handle to cover hard-to-reach areas
  • 1" bristle trim removes dirt from hard-to-reach crevices
  • Excellent for scraping off stains and dirt from metal surfaces
Equipped with a durable hardwood handle, these plater brushes can withstand the rigors of industrial usage.
Brass bristles: Ideal choice for plating or metal finishing in aircraft or missile industries, or other potentially flammable environments.
Tampico bristles: For general cleaning in the plating metal finishing aircraft and missile industries. Also used with inert arc aluminum welding
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty