Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Chipping Hammer Refill Brush, Weiler®
Supplier: Weiler
804-44070
300047-230EA 72.7 USD InStock
300047-230
Chipping Hammer Refill Brush, Weiler®
Brushes
Refill brush for most chipping hammers.
  • 0.0120" bristle diameter
  • Wood block
  • Steel bristles
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty