Would you like to log in again?
Econoline® Scratch Brushes, Weiler®
Supplier: Weiler
Econoline®
804-25201
300010-203EA 4.47 USD InStock
300010-203
Econoline® Scratch Brushes, Weiler®
Brushes
Brushes provide lasting, efficient service at minimal cost.
  • Sanded hardwood block finish
  • Slightly curved handle
For use cleaning dirt, rust, scale, chips, and paint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty