Would you like to log in again?
Valved 'Y' Connections, Western Enterprises
Supplier: Western Enterprises
312-111 312-411 312-112
300007-786EA 65.24 USD InStock
300007-786 300007-787 300007-849
Valved 'Y' Connections, Western Enterprises
Welding Supplies
Precision shut off valves
For use of two cutting and/or welding torches at once.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty