Would you like to log in again?
Hose Couplers, Western Enterprises
Supplier: Western Enterprises
312-C-31 312-230 312-231 312-D-30 312-D-31 312-C-30 312-31 312-131 312-130 312-30
300007-831EA 2.18 USD InStock
300007-831 300007-950 300066-758 300066-302 300007-791 300007-819 300066-688 300007-790 300066-760 300007-825
Hose Couplers, Western Enterprises
Welding Supplies
Made from brass to ensure functionality and durability.
Utilize hose couplers as a connector on the end of a hose to connect it with another hose or with a tap or a hose appliance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty