Would you like to log in again?
Pipe Thread Tees, Western Enterprises
Supplier: Western Enterprises
312-BFT-4SS 312-BFT-4HP 312-BST-4HP 312-BFT-4LP 312-BFT-6HP 312-BST-4LP 312-BMT-4HP
300066-658EA 50.8 USD InStock
300066-658 300066-646 300066-642 300007-948 300066-678 300066-644 300007-944
Pipe Thread Tees, Western Enterprises
Welding Supplies
  • Made from the highest quality material to ensure functionality and durability
  • Can withstand elevated pressures
  • Used to join poly pipe together
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty