Would you like to log in again?
Weiler® Burrite-Type Power Tube Brushes
Supplier: Weiler
Weiler®
804-21056 804-21055
300050-016CS 119.02 USD InStock
300050-016 300050-018
Weiler® Burrite-Type Power Tube Brushes
Brushes
Brushes are ideally suited for deburring, cleaning, and finishing applications.
  • Flat side action
  • For use with power drills and flexible shaft tools
Use with a holder on blind, threaded holes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty