Would you like to log in again?
Weiler® Vortec Pro® Turbo Tube™ Brushes
Supplier: Weiler
Vortec Pro®
804-36305
300085-636EA 84.85 USD InStock
300085-636
Weiler® Vortec Pro® Turbo Tube™ Brushes
Brushes
Cleans both I.D. and O.D. on fittings and copper tubing.
Two brushes in one, allowing users to clean fittings and copper tubing without changing tools. Plus, the tool is self-cleaning as it is switched from I.D. cleaning to O.D. cleaning, residual build-up is removed.
Diamater: 1" I.D. to 1 1/8" O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty