Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Weiler® 4-in-1 Tube and Fitting Brush
Supplier: Weiler
Weiler®
804-44690
300046-526EA 41.34 USD InStock
300046-526
Weiler® 4-in-1 Tube and Fitting Brush
Brushes
Cleans the outside and inside diameters of tubes and fittings.
Use with 3/4" and 1/2" tube I.D.s, and 7/8" and 5/8" tube O.D.s.
Ideally suited for cleaning, deburring, and surface preparation applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty