Would you like to log in again?
Industrial Torches, with Built-in Refillable Metal Fuel Tank and Removable Base, Master Appliance, ORS Nasco
Supplier: ORS Nasco
467-GT-70
76006-374EA 104.78 USD InStock
76006-374
Industrial Torches, with Built-in Refillable Metal Fuel Tank and Removable Base, Master Appliance, ORS Nasco
Soldering Supplies Soldering Irons
Use to braze with special alloy welding rods, heat and bend thin metals, loosen rusted bolts, pop dents out of metal, shrink and terminate heavy wall adhesive lined tubing, solder and desolder, and more.
  • 2500 °F operating temperature
  • Self-igniting piezo electric ignition is easy to start and eliminates the need for a lighter
  • Self-contained, metal fuel tank is rugged, lightweight, and easy to use, eliminating bulky canisters
  • Safety lock prevents unintentional ignition and locks for storage
  • 30 minute run time
  • Large flame accommodates heavy-duty"dustrial applications
Unit dimensions are 1.8" H x 6.5" L x 5" W
Meets CPSC requirements.
Butane gas required.
Certifications: Meets CPSC Requirements; CSA; TUV/GS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty