Would you like to log in again?
Williams Medium E, HiMedia Laboratories
Supplier: HiMedia Laboratories
AT125-1L AT125-10X1L AT125-5L AT125-20L
89238-986EA 31.65 USD InStock
89238-986 89238-988 89238-984 89238-990
Williams Medium E, HiMedia Laboratories
Cell Culture Media
William's medium E developed by Williams and Gunn for the isolation and long term maintenance of adult rat liver epithelial cells.
  • Medium contains L-glutamine and no sodium bicarbonate
  • Appearance: Off-white to creamish white, homogenous powder
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty