Would you like to log in again?
Instant-Release Attachments, MSA, ORS Nasco
Supplier: ORS Nasco
454-48733
300072-606EA 29.92 USD InStock
300072-606
Instant-Release Attachments, MSA, ORS Nasco
Tool Sets
Allows welding shields to be attached and detached quickly from headwear equipped with instant-release lugs.
Designed for use with MSA instant-release caps.
Delivery information: Includes screws and reinforcement plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty