Would you like to log in again?
Vortec Pro® Mounted Flap Wheels, Weiler
Supplier: Weiler
Vortec Pro®
804-30726 804-30720 804-30721 804-30728 804-30724
300045-334CS 35.92 USD InStock
300045-334 300045-336 300047-452 300046-304 300046-306
Vortec Pro® Mounted Flap Wheels, Weiler
Abrasives
  • A solid hub construction enables wheels to be rated at higher RPMs
  • Provides balance leading to smooth chatter-free finishing
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty