Would you like to log in again?
Lens Cleaning Towelettes, Anchor Products
Supplier: Anchor Brand
101-70-AB
300090-511EA 11.53 USD InStock
300090-511
Lens Cleaning Towelettes, Anchor Products
Wipes Disposable Wipes and Towels Lens Papers
Pre-moistened with C-Clear Anti-Fog, Anti-Static formula for use on plastic, polycarbonate, and glass lenses.
  • Contains no silicone
  • Dispenser packed
  • Safe and effective with all lens coatings, cluding Anti-Reflective coating
Strategic placement of towelette dispensers throughout a facility will promote proper lens cleaning habits and extend the life of prescription and non-prescription eyewear. Also good for use on goggles, face shields, CRT screens, and microscopes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty