Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SPC Dri-Zorb® Granular Absorbents, Brady
Supplier: Brady
655-DZ-100
300053-187EA 43.06 USD InStock
300053-187
SPC Dri-Zorb® Granular Absorbents, Brady
Spill Control Materials
An all-natural corncob product that quickly absorbs cutting oils, grease, water, and other liquids
Dri-Zorb® is particularly useful as an alternative to clay when disposal methods dictate the absorbent medium. Not for use with aggressive liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty