Would you like to log in again?
Slotted Shim Assortment Sets, ORS Nasco
Supplier: ORS Nasco
605-42910
300040-901EA 2174.15 USD InStock
300040-901
Slotted Shim Assortment Sets, ORS Nasco
Tool Sets
Slotted shims are the best way to solve alignment and leveling problems on motors, pumps, generators, and other plant machinery.
  • Marked with thickness in decimal and metric
  • 260 shims per set
  • 300 series hardness
Thicknesses are 0.0010", 0.0020", 0.0030", 0.0040", 0.0050", 0.0100", 0.0150", 0.0200", 0.0250", 0.0500", 0.0750", 0.1000", and 1/8". Includes 20 pieces of each thickness in a metal box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty