Would you like to log in again?
Proto® Super-Duty Cold Chisels, ORS Nasco
Supplier: ORS Nasco
577-86058 577-86078 577-86012 577-86001 577-86034 577-86038
300028-110EA 50.7 USD InStock
300028-110 300028-120 300028-118 300028-116 300028-114 300028-112
Proto® Super-Duty Cold Chisels, ORS Nasco
Chisels
Applicable for use with soft cast iron, brass, copper, aluminum, mild steel, bolts, and rivets.
  • Long knurled shank
  • Hand ground cutting edge
  • Hardened, quenched, and tempered to precise specifications
  • Black oxide finish
For use in extremely harsh environments and 20% larger striking surface than standard chisel.
Certifications: FED SPEC. GGG-C-313c; ANSI Spec. B209.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty