Would you like to log in again?
Proto® Cold Chisels, ORS Nasco
Supplier: ORS Nasco
577-86A1X8
300028-124EA 36.78 USD InStock
300028-124
Proto® Cold Chisels, ORS Nasco
Chisels
Applicable for use with soft cast iron, brass, copper, aluminum, mild steel, bolts, and rivets.
  • Hand ground cutting edge
  • Black oxide finish
Modified parabolic striking end to help reduce mushrooming and chipping.
Certifications: ANSI Spec. B209.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty