Would you like to log in again?
Proto® Super-Duty Cold Chisel Sets, ORS Nasco
Supplier: ORS Nasco
577-86C 577-86000 577-86B
300003-168PK 179.15 USD InStock
300003-168 300003-178 300003-180
Proto® Super-Duty Cold Chisel Sets, ORS Nasco
Chisels
Hand ground cutting edge.
  • Black oxide finish
  • Straight tips
  • English measurements
  • Packaged in kit pouch
Applicable for use with soft cast iron, copper, brass, aluminum, mild steel, bolts, and rivets.
Hardened, quenched, and tempered to precise specifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty