Would you like to log in again?
PVP Iodine Wipes, First Aid Only
Supplier: First Aid Only
G310-003
76049-188BX 17.19 USD InStock
76049-188
PVP Iodine Wipes, First Aid Only
First Aid Supplies First Aid Kit Components
These povidone iodine infection control wipes, in their convenient, single-use form, ward off infection when treating minor cuts, wounds, and abrasions.
  • Non-irritating and virtually non-stinging
  • Wipes measure 1.25×2.5"
  • Saturated with a 10% povidone-iodine solution
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty