Would you like to log in again?
SmartCompliance PVP Iodine Antiseptic Wipes Refill, First Aid Only
Supplier: First Aid Only
SmartCompliance
FAE-4005-001
76049-100BX 14.84 USD InStock
76049-100
SmartCompliance PVP Iodine Antiseptic Wipes Refill, First Aid Only
First Aid Supplies First Aid Kit Components
First Aid antiseptic and germicide to help prevent infection in minor cuts, scrapes, and minor burns.
  • Povidone Iodine is a broad-range antimicrobial that kills bacteria, fungi, viruses, protozoa, and yeast
  • 'Swab' style packaging offers exceptional shelf life
  • Contains Povidone Iodine (10% USP) antiseptic swabs, each containing 0.018 fluid oz. (0.5 cc) of medicated solution
Wipes are dispensed from the Smart Tab, ezRefill box, which includes an indicator tab to alert users when to reorder. The box snaps easily into place in all SmartCompliance cabinets, keeping supplies organized. For use with SmartCompliance systems: 90832, 90833, 90834, 90835.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty