Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proto® 440SS Tool Cabinets, 7 Drawers, Red, Stanley® Products

Supplier: Stanley
Proto®
577-442752-6RD 577-442742-7RD
300025-138EA 1555.65 USD InStock
300025-138 300025-134
Proto® 440SS Tool Cabinets, 7 Drawers, Red, Stanley® Products
Furniture Storage Cabinets
Steel construction with smooth, sleek and durable red paint finish.

  • Aluminum extruded drawer pulls
  • Recessed drawer pulls and 0.04" drawer front and body
  • Heavy duty key
  • Four-way channel bottom panel
  • 2" wide Polyurethane casters - 450 lbs/caster
  • Heavy duty easy grip handle

Maximum weight capacity for each drawer is 100 lbs.

Model 7 drawers: 2 drawers H17/8"; 4 drawers H315/16"; 1 drawer H81/16"
Model 6 drawers: 1 drawer H17/8"; 2 drawers H315/16"; 2 drawers H81/16" ; 1 drawer H121/8"

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty