Would you like to log in again?
STAT-8-Isoprostane ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
500431-96 500431-480
75816-798EA 363.88 USD InStock
75816-798 75816-796
STAT-8-Isoprostane ELISA Kit, Cayman Chemical
Assays ELISAs
For the rapid measurement of 8-iso PGF from biological samples.
  • Measure 8-isoprostane, a biomarker of oxidative stress
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 8-isoprostane levels down to 45 pg/mL
  • Extremely rapid assay; get results in about 2 hours
  • Alkaline phosphatase used as tracer
  • Midpoint: 50% B/B0: 100-300 pg/mL
Cayman’s STAT-8-isoprostane ELISA is a competitive assay that permits the rapid measurement of 8-iso PGF from biological samples, requiring only 1 hour incubation and development times for each step. This assay format is similar to that employed in Cayman’s original 8-isoprostane ELISA (Item No. 516351) with the only change being the use of an alkaline phosphatase tracer in place of an AChE tracer. While Item No. 516351 offers superior sensitivity (IC50 = ~35 pg/mL), the STAT-8-isoprostane assay offers the convenience of a fast assay while still achieving an IC50 value of 180 pg/mL and a detection limit (80% B/B0) of approximately 45 pg/mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty