Would you like to log in again?
Proto® Lockback Knives, Stanley® Products
Supplier: Stanley
577-18545
300024-120EA 115.28 USD InStock
300024-120
Proto® Lockback Knives, Stanley® Products
Knives
Knives with 420 grade high carbon stainless steel blade and hardwood handle with heavy-duty lock springs.
  • Solid one piece brass bolster and liner provide additional strength
  • Blade locks in open position to guard against accidental closing
  • J18545 includes a leather sheath
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty