Would you like to log in again?
Irwin® Bi-Metal Utility Knife Blades
Supplier: Irwin
586-2084100
300003-416PK 4.84 USD InStock
300003-416
Irwin® Bi-Metal Utility Knife Blades
Razors and Blades
High speed steel edge can be heat treated to higher hardness for longer life and edge sharpness.
  • Single cutting edge
  • Spring backing steel for blade flexibility and no dangerous shattering
  • Electron beam weld joins materials together
Stays sharper 3x longer than traditional carbon blades.
Packaging: 5 blades per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty